Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012614.1 | NP_001012632.1 | 4 | 663 | ||
NM_001328.2 | NP_001319.1 | 3 | 468 | |||
XM_005272261.3 | XP_005272318.1 | 3 | 479 | |||
XM_005272263.4 | XP_005272320.1 | 4 | 6694 | |||
XM_011513395.2 | XP_011511697.1 | 4 | 523 | |||
XM_011513396.2 | XP_011511698.1 | 4 | 628 | |||
XM_011513397.2 | XP_011511699.1 | 3 | 6411 | |||
XM_017007762.1 | XP_016863251.1 | 4 | 677 | |||
XM_017007763.1 | XP_016863252.1 | 4 | 800 | |||
XM_017007764.1 | XP_016863253.1 | 4 | 6694 | |||
XM_017007765.1 | XP_016863254.1 | 4 | 523 | |||
XM_017007766.1 | XP_016863255.1 | 4 | 628 | |||
XM_017007767.1 | XP_016863256.1 | 4 | 542 | |||
GenBank mRNA | AF091555.1 | - | 3 | 349 | ||
BC011655.2 | - | 3 | 341 | 4128336 | ||
BC053320.1 | - | 4 | 655 | 6461670 | ||
BC064333.1 | - | 4 | 2061 | |||
BC098400.1 | - | 3 | 478 | 30344174 | ||
KJ534818.1 | - | 3 | 303 | |||
KJ535006.1 | - | 3 | 303 | |||
KJ535050.1 | - | 2 | 148 | |||
KJ535065.1 | - | 4 | 498 | |||
KJ535066.1 | - | 4 | 498 | |||
KJ535067.1 | - | 4 | 498 | |||
U37408.1 | - | 3 | 352 |