Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 6 | 1184 | ||
XM_011520383.2 | XP_011518685.1 | 9 | 2110 | |||
XM_011520384.2 | XP_011518686.1 | 8 | 2221 | |||
XM_011520385.2 | XP_011518687.1 | 8 | 2224 | |||
XM_011520386.2 | XP_011518688.1 | 8 | 2224 | |||
XM_011520387.2 | XP_011518689.1 | 8 | 2224 | |||
XM_017018355.1 | XP_016873844.1 | 7 | 1562 | |||
GenBank mRNA | AK026288.1 | - | 3 | 595 | ||
AK074155.1 | - | 5 | 2050 | |||
AK090428.1 | - | 2 | 580 | |||
AK296137.1 | - | 2 | 63 | |||
BC066641.1 | - | 6 | 1104 | |||
BC109257.2 | - | 3 | 583 | 40024867 | ||
BC109258.2 | - | 3 | 583 | 40024874 |