Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025104.3 | NP_079380.2 | 3 | 353 | ||
NM_145663.2 | NP_663696.1 | 3 | 353 | |||
NR_036623.1 | 3 | 353 | ||||
XM_006722098.2 | XP_006722161.1 | 3 | 326 | |||
XM_006722102.3 | XP_006722165.1 | 2 | 316 | |||
XM_011525281.2 | XP_011523583.1 | 3 | 221 | |||
XM_017025145.1 | XP_016880634.1 | 3 | 326 | |||
XM_017025146.1 | XP_016880635.1 | 3 | 218 | |||
XM_017025147.1 | XP_016880636.1 | 2 | 317 | |||
XM_017025148.1 | XP_016880637.1 | 3 | 326 | |||
XM_017025149.1 | XP_016880638.1 | 3 | 326 | |||
XM_017025150.1 | XP_016880639.1 | 3 | 326 | |||
XM_017025151.1 | XP_016880640.1 | 3 | 326 | |||
XM_017025152.1 | XP_016880641.1 | 3 | 221 | |||
XM_017025153.1 | XP_016880642.1 | 3 | 216 | |||
XM_017025154.1 | XP_016880643.1 | 2 | 128 | |||
XM_017025155.1 | XP_016880644.1 | 2 | 316 | |||
XM_017025156.1 | XP_016880645.1 | 3 | 326 | |||
GenBank mRNA | AF448801.1 | - | 3 | 278 | ||
AK023149.1 | - | 3 | 305 | |||
AK295296.1 | - | 4 | 198 | |||
AK303282.1 | - | 3 | 325 | |||
BC016158.1 | - | 3 | 353 | 3536735 | ||
BC033660.2 | - | 3 | 275 | |||
DA475446.1 | - | 3 | 266 |