Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025108.2 | NP_079384.2 | 5 | 679 | ||
XM_011522667.1 | XP_011520969.1 | 4 | 583 | |||
GenBank mRNA | AK023971.1 | - | 4 | 760 | ||
AK093788.1 | - | 4 | 917 | |||
AK128408.1 | - | 5 | 1057 | |||
AK301929.1 | - | 5 | 675 | |||
AL559561.3 | - | 5 | 678 | |||
BC008882.2 | - | 2 | 331 | |||
BC018719.1 | - | 5 | 667 | 4868952 | ||
CR457319.1 | - | 2 | 147 |