Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 6 | 1485 | ||
NM_001286762.1 | NP_001273691.1 | 5 | 1391 | |||
NM_001286763.1 | NP_001273692.1 | 6 | 1120 | |||
NM_001286764.1 | NP_001273693.1 | 5 | 1004 | |||
NM_001286765.1 | NP_001273694.1 | 5 | 850 | |||
NM_001286766.1 | NP_001273695.1 | 4 | 734 | |||
NM_025113.3 | NP_079389.2 | 6 | 1507 | |||
GenBank mRNA | AK025215.1 | - | 5 | 1140 | ||
AK093073.1 | - | 6 | 1455 | |||
AK125950.1 | - | 6 | 1492 | |||
AK293034.1 | - | 5 | 756 | |||
AK300183.1 | - | 5 | 554 | |||
AK307854.1 | - | 4 | 477 | |||
AK310264.1 | - | 7 | 1494 | |||
BC004495.1 | - | 2 | 204 | |||
BC032311.1 | - | 5 | 1072 | |||
BC043488.1 | - | 6 | 853 | 5205680 | ||
BX641126.1 | - | 4 | 541 |