Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078650.2 | NP_001072118.1 | 5, 6 | 547 | ||
NM_001078651.2 | NP_001072119.1 | 6, 7 | 565 | |||
NM_025124.3 | NP_079400.1 | 6, 7 | 592 | |||
NR_073409.1 | 6, 7 | 759 | ||||
NR_073410.1 | 5, 6 | 531 | ||||
NR_073411.1 | 6, 7 | 603 | ||||
NR_073413.1 | 6, 7 | 576 | ||||
XM_006718693.2 | XP_006718756.1 | 5, 6 | 515 | |||
GenBank mRNA | AK025402.1 | - | 6, 7 | 608 | ||
BC013883.1 | - | 6, 7 | 728 | 3678954 | ||
BC125134.1 | - | 5, 6 | 515 | 40119367 | ||
BG488634.1 | - | 6, 7 | 514 | |||
BG719036.1 | - | 5, 6 | 527 | |||
BM920758.1 | - | 560 | ||||
CN336445.1 | - | 6, 7 | 426 | |||
DN999439.1 | - | 5, 6 | 536 | |||
HY059123.1 | - | 6, 7 | 519 |