Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256858.1 | NP_001243787.1 | 2 | 150 | ||
NM_025126.3 | NP_079402.2 | 3 | 726 | |||
NM_194271.2 | NP_919247.1 | 4 | 757 | |||
GenBank mRNA | AB084914.1 | - | 2 | 599 | ||
AF306709.1 | - | 3 | 768 | |||
AK025439.1 | - | 3 | 691 | |||
AK225285.1 | - | 3 | 691 | |||
AK304366.1 | - | 4 | 746 | |||
AL557811.3 | - | 4 | 767 | |||
AY098934.1 | - | 2 | 596 | |||
BC007826.2 | - | 3 | 651 | 3347885 | ||
BI559962.1 | - | 2 | 125 | |||
BT007283.1 | - | 2 | 596 | |||
CR457342.1 | - | 2 | 596 |