Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190274.1 | NP_001177203.1 | 15 | 1966 | ||
NM_025133.4 | NP_079409.3 | 15 | 1755 | |||
XM_005264572.4 | XP_005264629.1 | 15 | 1960 | |||
XM_005264573.4 | XP_005264630.1 | 15 | 1913 | |||
XM_017005015.1 | XP_016860504.1 | 15 | 1913 | |||
XM_017005016.1 | XP_016860505.1 | 15 | 1779 | |||
XM_017005017.1 | XP_016860506.1 | 15 | 1779 | |||
GenBank mRNA | AF351618.1 | - | 15 | 1768 | ||
AK022735.1 | - | 11 | 1268 | |||
AK097734.1 | - | 4 | 503 | |||
AK292877.1 | - | 15 | 1805 | |||
AK304140.1 | - | 15 | 1670 | |||
AK315108.1 | - | 15 | 1755 | |||
AY827075.1 | - | 15 | 1893 | |||
BC012728.2 | - | 1 | 39 | |||
BC043258.1 | - | 15 | 1857 | |||
BC130445.1 | - | 15 | 1691 | 40146476 | ||
BC136480.1 | - | 15 | 1691 | 9020469 |