Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142544.1 | NP_001136016.1 | 3 | 745 | ||
NM_001330.3 | NP_001321.1 | 3 | 748 | |||
XM_011545759.2 | XP_011544061.1 | 3 | 857 | |||
XM_011545760.2 | XP_011544062.1 | 3 | 815 | |||
GenBank mRNA | BC036787.1 | - | 3 | 714 | 5754188 | |
BC049822.1 | - | 3 | 732 | |||
BC064416.1 | - | 3 | 748 | 5750481 | ||
U43030.1 | - | 3 | 743 |