Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD9 SIRNA Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
UniGene ID:
Hs.59159
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37052
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHD9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308319.1
NP_001295248.1
11
2955
NM_025134.4
NP_079410.4
12
3000
XM_005256168.3
XP_005256225.1
12
3042
XM_005256169.2
XP_005256226.1
12
3064
XM_005256172.3
XP_005256229.1
12
2981
XM_005256176.3
XP_005256233.1
11
1649
XM_006721281.2
XP_006721344.1
12
2981
XM_006721283.3
XP_006721346.1
12
3202
XM_011523347.2
XP_011521649.1
13
3114
XM_017023718.1
XP_016879207.1
13
3237
XM_017023719.1
XP_016879208.1
14
3188
XM_017023720.1
XP_016879209.1
13
3082
XM_017023721.1
XP_016879210.1
13
3238
XM_017023722.1
XP_016879211.1
13
3238
XM_017023723.1
XP_016879212.1
13
3238
XM_017023724.1
XP_016879213.1
13
3064
XM_017023725.1
XP_016879214.1
13
3064
XM_017023726.1
XP_016879215.1
11
2271
XM_017023727.1
XP_016879216.1
11
1649
XM_017023728.1
XP_016879217.1
13
3098
GenBank mRNA
AB002306.3
-
11
2423
AK022240.1
-
2
328
AK022582.1
-
1
138
AK299045.1
-
11
1698
AY243500.1
-
11
2878
AY647157.1
-
12
3000
BC140815.1
-
11
2832
9021683
BC144622.1
-
11
2832
9053153
DQ059482.1
-
2
328
DQ333316.1
-
12
3094
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline