Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308445.1 | NP_001295374.1 | 3, 4 | 605 | ||
NM_025146.3 | NP_079422.1 | 3, 4 | 608 | |||
GenBank mRNA | AK023090.1 | - | 3, 4 | 523 | ||
AK023256.1 | - | 3, 4 | 558 | |||
AK296456.1 | - | 3, 4 | 303 | |||
AK307184.1 | - | 4, 5 | 452 | |||
AK315876.1 | - | 3, 4 | 410 | |||
AL601126.1 | - | 3, 4 | 503 | |||
BC012731.2 | - | 3, 4 | 547 | 3619903 |