Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271895.1 | NP_001258824.1 | 8 | 910 | ||
NM_001271896.1 | NP_001258825.1 | 6 | 766 | |||
NM_025150.4 | NP_079426.2 | 10 | 1156 | |||
NR_073513.1 | 6 | 555 | ||||
NR_073514.1 | 9 | 1031 | ||||
XM_006711555.1 | XP_006711618.1 | 10 | 1119 | |||
XM_006711556.1 | XP_006711619.1 | 9 | 972 | |||
XM_017002394.1 | XP_016857883.1 | 9 | 972 | |||
XM_017002395.1 | XP_016857884.1 | 8 | 873 | |||
GenBank mRNA | AK022590.1 | - | 6 | 716 | ||
AK222814.1 | - | 10 | 1097 | |||
AK225368.1 | - | 8 | 830 | |||
AK298773.1 | - | 9 | 984 | |||
AK315507.1 | - | 8 | 856 | |||
AY450558.1 | - | 6 | 485 | |||
BC000541.2 | - | 10 | 1103 | 3163506 | ||
BC007824.2 | - | 10 | 1044 | |||
BC009997.2 | - | 10 | 1076 | 4128881 | ||
BP308698.1 | - | 2 | 73 |