Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025152.2 | NP_079428.2 | 3 | 329 | ||
NR_120408.1 | 3 | 329 | ||||
XM_011537182.2 | XP_011535484.1 | 3 | 238 | |||
XM_011537183.2 | XP_011535485.1 | 3 | 315 | |||
XM_017021664.1 | XP_016877153.1 | 3 | 315 | |||
XM_017021665.1 | XP_016877154.1 | 3 | 315 | |||
XM_017021666.1 | XP_016877155.1 | 3 | 315 | |||
XM_017021667.1 | XP_016877156.1 | 3 | 290 | |||
GenBank mRNA | AK022722.1 | - | 3 | 242 | ||
AK295326.1 | - | 3 | 290 | |||
BC024919.1 | - | 3 | 255 | 4475196 | ||
BP274976.1 | - | 3 | 329 | |||
BX248028.1 | - | 3 | 316 | |||
KJ534907.1 | - | 3 | 274 | |||
KJ534908.1 | - | 3 | 274 | |||
KJ534909.1 | - | 3 | 274 |