Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025163.3 | NP_079439.2 | 3 | 1079 | ||
XM_011513190.1 | XP_011511492.1 | 4 | 1670 | |||
XM_011513192.2 | XP_011511494.1 | 4 | 1196 | |||
XM_017007240.1 | XP_016862729.1 | 3 | 2037 | |||
XM_017007241.1 | XP_016862730.1 | 4 | 3414 | |||
XM_017007242.1 | XP_016862731.1 | 3 | 3106 | |||
XM_017007243.1 | XP_016862732.1 | 3 | 998 |