Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025163.3 | NP_079439.2 | 2 | 267 | ||
XM_011513190.1 | XP_011511492.1 | 3 | 858 | |||
XM_011513192.2 | XP_011511494.1 | 3 | 384 | |||
XM_017007240.1 | XP_016862729.1 | 2 | 1225 | |||
XM_017007241.1 | XP_016862730.1 | 3 | 2602 | |||
XM_017007242.1 | XP_016862731.1 | 2 | 807 | |||
GenBank mRNA | AK022830.1 | - | 2 | 229 | ||
BC018804.1 | - | 2 | 148 | |||
BC044640.1 | - | 2 | 149 | 4871581 | ||
BG760692.1 | - | 2 | 131 | |||
DA736224.1 | - | 2 | 267 |