Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTNNA1 SIRNA Assays ›
Gene Symbol:
CTNNA1
Gene Name:
catenin alpha 1
Gene Aliases:
CAP102, MDPT2
Chromosome Location:
Chr.5: 138753386 - 138935034 on Build GRCh38
UniGene ID:
Hs.445981
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s3717
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTNNA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001290307.2
NP_001277236.1
10
1472
NM_001290309.2
NP_001277238.1
9
1276
NM_001290310.2
NP_001277239.1
10
1309
NM_001290312.1
NP_001277241.1
4
792
NM_001323982.1
NP_001310911.1
11
1942
NM_001323983.1
NP_001310912.1
10
1624
NM_001323984.1
NP_001310913.1
11
1490
NM_001323985.1
NP_001310914.1
10
1472
NM_001323987.1
NP_001310916.1
4
481
NM_001323988.1
NP_001310917.1
7
729
NM_001323989.1
NP_001310918.1
5
577
NM_001323990.1
NP_001310919.1
6
941
NM_001323991.1
NP_001310920.1
5
888
NM_001323992.1
NP_001310921.1
7
721
NM_001323993.1
NP_001310922.1
7
1035
NM_001323994.1
NP_001310923.1
4
490
NM_001323995.1
NP_001310924.1
7
705
NM_001323996.1
NP_001310925.1
7
1040
NM_001323997.1
NP_001310926.1
7
724
NM_001323998.1
NP_001310927.1
7
1016
NM_001323999.1
NP_001310928.1
7
1013
NM_001324000.1
NP_001310929.1
5
885
NM_001324001.1
NP_001310930.1
4
792
NM_001324002.1
NP_001310931.1
4
792
NM_001324003.1
NP_001310932.1
6
941
NM_001324004.1
NP_001310933.1
4
481
NM_001324005.1
NP_001310934.1
7
1016
NM_001324006.1
NP_001310935.1
7
705
NM_001324007.1
NP_001310936.1
6
941
NM_001324008.1
NP_001310937.1
4
481
NM_001324009.1
NP_001310938.1
4
792
NM_001324012.1
NP_001310941.1
2
247
NM_001324013.1
NP_001310942.1
5
821
NM_001903.4
NP_001894.2
10
1472
GenBank mRNA
AK094151.1
-
7
681
AK296708.1
-
10
1280
AK300439.1
-
9
1243
AK304267.1
-
10
1367
BC000385.2
-
10
1410
2821111
BC031262.1
-
4
371
5274351
D13866.1
-
10
1411
D14705.1
-
10
1436
HQ589335.1
-
4
626
L23805.1
-
9
1349
U03100.1
-
9
1349
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
actin filament organization
cell adhesion
establishment or maintenance of cell polarity
negative regulation of neuroblast proliferation
aging
male gonad development
gap junction assembly
axon regeneration
adherens junction organization
cellular protein localization
odontogenesis of dentin-containing tooth
apical junction assembly
response to estrogen
positive regulation of smoothened signaling pathway
positive regulation of muscle cell differentiation
protein heterooligomerization
cellular response to indole-3-methanol
epithelial cell-cell adhesion
negative regulation of cell motility
negative regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
structural molecule activity
protein binding
beta-catenin binding
vinculin binding
poly(A) RNA binding
gamma-catenin binding
cadherin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline