Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320484.1 | NP_001307413.1 | 10 | 1411 | ||
NM_001320485.1 | NP_001307414.1 | 10 | 1399 | |||
NM_001320487.1 | NP_001307416.1 | 10 | 1433 | |||
NM_025204.3 | NP_079480.2 | 10 | 1424 | |||
NR_135275.1 | 10 | 1395 | ||||
XM_011530713.2 | XP_011529015.2 | 10 | 1469 | |||
XM_017028944.1 | XP_016884433.1 | 10 | 1511 | |||
GenBank mRNA | AF218001.1 | - | 2 | 1638 | ||
AK024447.1 | - | 1 | 4580 | |||
AL449244.1 | - | 10 | 1405 | |||
AY203945.1 | - | 9 | 1471 | |||
BC012445.1 | - | 10 | 1243 | 3857552 | ||
BC093029.1 | - | 9 | 1497 | 30527560 |