Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030624.2 | NP_085127.2 | 3 | 542 | ||
XM_006724518.1 | XP_006724581.1 | 2 | 754 | |||
GenBank mRNA | AB051464.1 | - | 1 | 39 | ||
AK057298.1 | - | 3 | 542 | |||
BC109058.2 | - | 1 | 286 | 40033682 | ||
BC109059.2 | - | 1 | 286 | 40033683 | ||
EU794630.1 | - | 1 | 286 |