Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272004.1 | NP_001258933.1 | 10 | 1855 | ||
NM_001272019.2 | NP_001258948.1 | 10 | 1461 | |||
NM_001282391.1 | NP_001269320.1 | 10 | 1707 | |||
NM_025209.3 | NP_079485.1 | 10 | 1855 | |||
XM_006717510.1 | XP_006717573.1 | 10 | 1707 | |||
GenBank mRNA | AF277374.1 | - | 10 | 1822 | ||
AF286905.1 | - | 10 | 1525 | |||
AK024329.1 | - | 7 | 970 | |||
AK092304.1 | - | 10 | 1461 | |||
AK299676.1 | - | 10 | 1648 | |||
AK309309.1 | - | 10 | 1593 | |||
AK309822.1 | - | 10 | 1724 | |||
BC036529.1 | - | 10 | 1856 | 4828057 | ||
CR749375.1 | - | 8 | 4449 |