Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164883.1 | NP_001158355.1 | 15 | 2405 | ||
NM_001282597.2 | NP_001269526.1 | 15 | 2405 | |||
NM_001282598.1 | NP_001269527.1 | 15 | 2258 | |||
NM_001282599.1 | NP_001269528.1 | 9 | 1346 | |||
NM_001282600.1 | NP_001269529.1 | 9 | 1289 | |||
NM_001320810.1 | NP_001307739.1 | 9 | 1289 | |||
NM_004389.3 | NP_004380.2 | 15 | 2405 | |||
XM_011532555.2 | XP_011530857.1 | 15 | 2252 | |||
XM_011532556.2 | XP_011530858.1 | 16 | 2448 | |||
XM_011532557.2 | XP_011530859.1 | 7 | 994 | |||
XM_017003403.1 | XP_016858892.1 | 16 | 2448 | |||
XM_017003404.1 | XP_016858893.1 | 15 | 2343 | |||
XM_017003405.1 | XP_016858894.1 | 15 | 2343 | |||
XM_017003406.1 | XP_016858895.1 | 9 | 1379 | |||
GenBank mRNA | AK127226.1 | - | 19 | 2851 | ||
AK294362.1 | - | 1 | 370 | |||
AK295181.1 | - | 15 | 2204 | |||
AK295493.1 | - | 9 | 1276 | |||
AK303035.1 | - | 15 | 2258 | |||
BC040458.1 | - | 14 | 2077 | 4815068 | ||
BC052996.1 | - | 15 | 2339 | 6187158 | ||
BX537769.1 | - | 9 | 1346 | |||
BX648368.1 | - | 2 | 218 | |||
HQ589336.1 | - | 9 | 1277 | |||
M94151.2 | - | 14 | 2156 |