Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258248.1 | NP_001245177.1 | 2 | 374 | ||
NM_199262.2 | NP_954871.1 | 2 | 363 | |||
XM_006722115.3 | XP_006722178.1 | 2 | 326 | |||
GenBank mRNA | AK127707.1 | - | 2 | 417 | ||
AK127850.1 | - | 2 | 363 | |||
BC103951.1 | - | 2 | 262 | 40011121 | ||
BC103952.1 | - | 2 | 262 | 40011125 | ||
BC103953.1 | - | 2 | 262 | 40011129 | ||
BC103954.1 | - | 2 | 262 | 40011132 |