Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032548.3 | NP_115937.1 | 12 | 1963 | ||
NM_172027.2 | NP_742024.1 | 12 | 1952 | |||
NR_033429.1 | 12 | 1917 | ||||
XM_006713769.3 | XP_006713832.1 | 12 | 1986 | |||
XM_011513213.1 | XP_011511515.1 | 12 | 1886 | |||
XM_017007285.1 | XP_016862774.1 | 11 | 1815 | |||
XM_017007286.1 | XP_016862775.1 | 11 | 1797 | |||
XM_017007287.1 | XP_016862776.1 | 12 | 1923 | |||
XM_017007288.1 | XP_016862777.1 | 11 | 1856 | |||
GenBank mRNA | AB053324.1 | - | 12 | 1942 | ||
AB053325.1 | - | 12 | 1931 | |||
AB053326.1 | - | 12 | 1896 | |||
AF447886.1 | - | 11 | 2056 | |||
AK098738.1 | - | 5 | 1083 | |||
AK130594.1 | - | 9 | 1369 | |||
BC011858.2 | - | 12 | 1950 | 4309664 |