Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303247.1 | NP_001290176.1 | 8, 9 | 963 | ||
NM_001303248.1 | NP_001290177.1 | 6, 7 | 578 | |||
NM_025234.2 | NP_079510.1 | 8, 9 | 857 | |||
XM_011522094.1 | XP_011520396.1 | 8, 9 | 837 | |||
XM_017022637.1 | XP_016878126.1 | 8, 9 | 753 | |||
GenBank mRNA | AF100786.1 | - | 8, 9 | 718 | ||
AF127799.1 | - | 7, 8 | 706 | |||
AF309553.1 | - | 7, 8 | 702 | |||
AK024754.1 | - | 8, 9 | 729 | |||
BC010080.2 | - | 8, 9 | 721 | 4329599 | ||
BU528743.1 | - | 6, 7 | 443 | |||
CR457333.1 | - | 6, 7 | 653 |