Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025236.3 | NP_079512.2 | 4 | 1508 | ||
NM_170769.2 | NP_739575.2 | 5 | 1308 | |||
XM_017011325.1 | XP_016866814.1 | 3 | 1071 | |||
GenBank mRNA | AF238315.1 | - | 4 | 1509 | ||
AF238316.1 | - | 5 | 1308 | |||
AF238317.1 | - | 2 | 674 |