Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000291.3 | NP_000282.1 | 4, 5 | 590 | ||
GenBank mRNA | AB062432.1 | - | 4, 5 | 487 | ||
AK291081.1 | - | 4, 5 | 504 | |||
AK295182.1 | - | 4, 5 | 394 | |||
AK298855.1 | - | 470 | ||||
AK301632.1 | - | 4, 5 | 503 | |||
AK301740.1 | - | 4, 5 | 663 | |||
AK303848.1 | - | 3, 4 | 243 | |||
AK310756.1 | - | 4, 5 | 1165 | |||
AK312280.1 | - | 4, 5 | 504 | |||
AY423725.1 | - | 4, 5 | 440 | |||
BC023234.1 | - | 4, 5 | 454 | 3917985 | ||
BC103752.1 | - | 4, 5 | 504 | 6526214 | ||
BC104837.1 | - | 4, 5 | 469 | 8143840 | ||
BC113568.1 | - | 4, 5 | 469 | 8322620 | ||
CR456716.1 | - | 4, 5 | 418 | |||
EU794645.1 | - | 4, 5 | 504 | |||
L00160.1 | - | 3, 4 | 353 | |||
V00572.1 | - | 4, 5 | 497 |