Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025245.2 | NP_079521.1 | 6 | 1086 | ||
NR_038198.1 | 6 | 985 | ||||
XM_006722911.3 | XP_006722974.1 | 5 | 667 | |||
XM_011528320.2 | XP_011526622.1 | 7 | 1132 | |||
XM_011528322.2 | XP_011526624.1 | 5 | 721 | |||
XM_017027326.1 | XP_016882815.1 | 7 | 767 | |||
XM_017027327.1 | XP_016882816.1 | 7 | 748 | |||
XM_017027328.1 | XP_016882817.1 | 7 | 1132 | |||
XM_017027329.1 | XP_016882818.1 | 4 | 557 | |||
GenBank mRNA | AJ300182.1 | - | 5 | 667 | ||
AK097427.1 | - | 6 | 823 | |||
AK301631.1 | - | 6 | 750 | |||
BC141859.1 | - | 6 | 1086 | 8860384 | ||
BC143588.1 | - | 6 | 761 | |||
BC143589.1 | - | 6 | 772 |