Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136538.1 | NP_001130010.1 | 7 | 1086 | ||
NM_025247.5 | NP_079523.3 | 6 | 993 | |||
GenBank mRNA | AK092356.1 | - | 6 | 978 | ||
AL832043.1 | - | 7 | 1064 | |||
AY323912.1 | - | 6 | 949 | |||
BC003698.1 | - | 2 | 150 | |||
BC015056.1 | - | 6 | 965 | |||
BC126358.1 | - | 6 | 907 | 8992074 |