Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136538.1 | NP_001130010.1 | 8 | 1213 | ||
NM_025247.5 | NP_079523.3 | 7 | 1120 | |||
GenBank mRNA | AK092356.1 | - | 7 | 1105 | ||
AK097425.1 | - | 2 | 252 | |||
AL832043.1 | - | 8 | 1191 | |||
AY323912.1 | - | 7 | 1076 | |||
BC003698.1 | - | 3 | 277 | |||
BC126358.1 | - | 7 | 1034 | 8992074 |