Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025248.2 | NP_079524.2 | 19 | 3736 | ||
XM_017025169.1 | XP_016880658.1 | 23 | 4828 | |||
XM_017025170.1 | XP_016880659.1 | 21 | 4026 | |||
XM_017025171.1 | XP_016880660.1 | 20 | 3903 | |||
XM_017025172.1 | XP_016880661.1 | 19 | 3801 | |||
GenBank mRNA | AB051471.1 | - | 13 | 2248 | ||
AB127405.1 | - | 19 | 3736 |