Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005385.1 | - | 5 | 391 | 3950220 | |
BT007356.1 | - | 4 | 349 | |||
BX497259.1 | - | 5 | 370 | |||
CA946996.1 | - | 4 | 310 | |||
M24400.1 | - | 4 | 365 |
Gene Symbol: CTRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025200.3 | NP_001020371.3 | 5 | 382 | ||
GenBank mRNA | AK124403.1 | - | 3 | 1355 | ||
AK131056.1 | - | 5 | 369 | |||
AK225933.1 | - | 5 | 369 | |||
AK291822.1 | - | 5 | 367 | |||
BC073145.1 | - | 5 | 358 | 5747536 | ||
BP324399.1 | - | 5 | 382 |