Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THUMPD2 SIRNA Assays ›
Gene Symbol:
THUMPD2
Gene Name:
THUMP domain containing 2
Gene Aliases:
C2orf8
Chromosome Location:
Chr.2: 39736060 - 39779276 on Build GRCh38
UniGene ID:
Hs.468254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37346
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THUMPD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321468.1
NP_001308397.1
4, 5
784
NM_001321469.1
NP_001308398.1
4, 5
774
NM_001321470.1
NP_001308399.1
4, 5
726
NM_001321474.1
NP_001308403.1
4, 5
784
NM_001321475.1
NP_001308404.1
4, 5
774
NM_001321477.1
NP_001308406.1
5, 6
967
NM_001321478.1
NP_001308407.1
5, 6
1075
NM_001321479.1
NP_001308408.1
3, 4
374
NM_001321480.1
NP_001308409.1
5, 6
1065
NM_001321481.1
NP_001308410.1
5, 6
977
NM_025264.4
NP_079540.2
4, 5
784
NR_028102.1
4, 5
784
NR_135659.1
5, 6
1075
NR_135660.1
5, 6
977
NR_135661.1
5, 6
1065
NR_135662.1
4, 5
774
NR_135663.1
3, 4
374
NR_135664.1
5, 6
967
XM_017005050.1
XP_016860539.1
4, 5
736
XM_017005051.1
XP_016860540.1
4, 5
736
XM_017005054.1
XP_016860543.1
4, 5
1465
GenBank mRNA
AF380577.1
-
1, 2
80
AF380578.1
-
1, 2
1588
AF380579.1
-
1, 2
754
AK057801.1
-
2, 3
166
AK093580.1
-
4, 5
755
AK226034.1
-
4, 5
736
AK292002.1
-
4, 5
769
AK298178.1
-
4, 5
671
AK301353.1
-
5, 6
853
AL832115.1
-
4, 5
751
BC004163.2
-
4, 5
745
2961526
BC013299.2
-
4, 5
745
2961526
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
tRNA methylation
tRNA binding
tRNA (guanine) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline