Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030572.3 | NP_085049.1 | 2, 3 | 309 | ||
GenBank mRNA | AK027273.1 | - | 2, 3 | 147 | ||
AK075342.1 | - | 2, 3 | 263 | |||
BC004336.1 | - | 2, 3 | 244 | 3631700 | ||
DA380664.1 | - | 2, 3 | 308 |