Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181900.2 | NP_871629.1 | 3 | 318 | ||
NR_135013.1 | 2 | 268 | ||||
XM_006720703.1 | XP_006720766.1 | 3 | 475 | |||
XM_017022639.1 | XP_016878128.1 | 2 | 833 | |||
GenBank mRNA | AF480304.1 | - | 3 | 234 | ||
AK026352.1 | - | 2 | 226 | |||
AK097024.1 | - | 1 | 1686 | |||
AK225479.1 | - | 2 | 226 | |||
AL137657.1 | - | 2 | 268 | |||
BC004365.1 | - | 3 | 260 | 3641640 |