Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029885.1 | NP_001025056.1 | 3 | 1194 | ||
XM_005244801.3 | XP_005244858.1 | 3 | 1307 | |||
XM_005244802.1 | XP_005244859.1 | 2 | 1004 | |||
XM_011542200.2 | XP_011540502.1 | 3 | 1203 | |||
GenBank mRNA | AL122073.1 | - | 1 | 302 | ||
BC004366.1 | - | 1 | 205 | |||
BC098429.1 | - | 1 | 618 |