Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029885.1 | NP_001025056.1 | 2 | 480 | ||
XM_005244801.3 | XP_005244858.1 | 2 | 593 | |||
XM_011542200.2 | XP_011540502.1 | 2 | 489 | |||
GenBank mRNA | JN542538.1 | - | 1 | 65 |