Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317237.1 | NP_001304166.1 | 6 | 629 | ||
NM_001908.4 | NP_001899.1 | 6 | 660 | |||
NM_147780.3 | NP_680090.1 | 8 | 822 | |||
NM_147781.3 | NP_680091.1 | 7 | 779 | |||
NM_147782.3 | NP_680092.1 | 7 | 748 | |||
NM_147783.3 | NP_680093.1 | 7 | 734 | |||
XM_006716244.2 | XP_006716307.1 | 8 | 846 | |||
XM_006716245.2 | XP_006716308.1 | 6 | 950 | |||
XM_011543812.2 | XP_011542114.1 | 8 | 755 | |||
XM_017013097.1 | XP_016868586.1 | 7 | 736 | |||
XM_017013098.1 | XP_016868587.1 | 7 | 736 | |||
XM_017013099.1 | XP_016868588.1 | 8 | 778 | |||
XM_017013100.1 | XP_016868589.1 | 7 | 680 | |||
XM_017013101.1 | XP_016868590.1 | 3 | 203 | |||
GenBank mRNA | AK075393.1 | - | 6 | 636 | ||
AK092070.1 | - | 7 | 671 | |||
AK097384.1 | - | 1 | 590 | |||
AK130184.1 | - | 6 | 597 | |||
AK290239.1 | - | 6 | 597 | |||
AK297687.1 | - | 4 | 361 | |||
BC010240.1 | - | 7 | 677 | 4154307 | ||
BC095408.1 | - | 6 | 597 | 30334082 | ||
BM008741.1 | - | 6 | 639 | |||
BX647765.1 | - | 6 | 520 | |||
BX647952.1 | - | 6 | 600 | |||
BX648239.1 | - | 3 | 4671 | |||
L16510.1 | - | 7 | 662 | |||
M13230.1 | - | 2 | 95 | |||
M14221.1 | - | 7 | 679 | |||
Y18460.1 | - | 2 | 167 |