Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030579.2 | NP_085056.2 | 2, 3 | 394 | ||
GenBank mRNA | AB009282.1 | - | 2, 3 | 421 | ||
AK291576.1 | - | 2, 3 | 326 | |||
BC004373.1 | - | 2, 3 | 329 | 3639133 | ||
BC014431.1 | - | 2, 3 | 352 | 3542294 | ||
BX647922.1 | - | 2, 3 | 370 |