Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286769.1 | NP_001273698.1 | 6 | 1327 | ||
NM_001286770.1 | NP_001273699.1 | 5 | 1203 | |||
NM_030580.4 | NP_085057.3 | 6 | 1323 | |||
XM_011517314.2 | XP_011515616.1 | 6 | 2650 | |||
XM_011517315.2 | XP_011515617.1 | 6 | 1567 | |||
XM_011517316.2 | XP_011515618.1 | 7 | 1372 | |||
XM_011517318.2 | XP_011515620.1 | 6 | 2650 | |||
XM_011517320.2 | XP_011515622.1 | 5 | 1233 | |||
XM_017013872.1 | XP_016869361.1 | 7 | 1373 | |||
XM_017013873.1 | XP_016869362.1 | 6 | 1313 | |||
XM_017013874.1 | XP_016869363.1 | 6 | 1347 | |||
XM_017013875.1 | XP_016869364.1 | 7 | 1513 | |||
XM_017013876.1 | XP_016869365.1 | 5 | 1187 | |||
GenBank mRNA | AK096508.1 | - | 5 | 1174 | ||
AL833814.1 | - | 6 | 1307 | |||
BC004480.2 | - | 6 | 1282 | 3938462 | ||
BC028136.1 | - | 3 | 1425 | 5215496 |