Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 33 | 3314 | ||
NM_130444.2 | NP_569711.2 | 33 | 4019 | |||
NM_130445.3 | NP_569712.2 | 34 | 2847 | |||
GenBank mRNA | AF018081.1 | - | 33 | 3333 | ||
AF018082.1 | - | 34 | 2812 | |||
BC033715.1 | - | 22 | 1195 | |||
BC063833.1 | - | 34 | 2788 | 6181818 | ||
L22548.1 | - | 15 | 794 |