Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 28 | 3011 | ||
NM_130444.2 | NP_569711.2 | 28 | 3716 | |||
NM_130445.3 | NP_569712.2 | 29 | 2544 | |||
GenBank mRNA | AF018081.1 | - | 28 | 3030 | ||
AF018082.1 | - | 29 | 2509 | |||
BC033715.1 | - | 17 | 892 | |||
BC063833.1 | - | 29 | 2485 | 6181818 | ||
L22548.1 | - | 10 | 491 |