Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030628.1 | NP_085131.1 | 2 | 1741 | ||
GenBank mRNA | AB051485.1 | - | 1 | 1355 | ||
AK074261.1 | - | 1 | 959 | |||
AK123587.1 | - | 1 | 1583 | |||
BC028025.1 | - | 1 | 1130 | |||
BC060841.1 | - | 2 | 1741 | 30338563 |