Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142524.1 | NP_001135996.1 | 4 | 1194 | ||
NM_001142525.1 | NP_001135997.1 | 4 | 1190 | |||
NM_001142526.1 | NP_001135998.1 | 4 | 1126 | |||
NM_001142527.1 | NP_001135999.1 | 4 | 1178 | |||
NM_001142528.1 | NP_001136000.1 | 4 | 1110 | |||
NM_001142529.1 | NP_001136001.1 | 3 | 1143 | |||
NM_001142530.1 | NP_001136002.1 | 2 | 1048 | |||
NM_030639.2 | NP_085142.1 | 3 | 1095 | |||
GenBank mRNA | AB051488.1 | - | 2 | 938 | ||
AF547055.1 | - | 1 | 558 | |||
BC041409.2 | - | 3 | 1083 | 5284828 |