Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138697.3 | NP_619642.2 | 1, 2 | 295 | ||
NM_177540.2 | NP_803884.1 | 1, 2 | 295 | |||
XM_011542206.2 | XP_011540508.1 | 1 | 847 | |||
XM_017002402.1 | XP_016857891.1 | 1 | 847 | |||
XM_017002403.1 | XP_016857892.1 | 1 | 847 | |||
GenBank mRNA | AK292014.1 | - | 1, 2 | 264 | ||
AK293499.1 | - | 1, 2 | 164 | |||
AK315805.1 | - | 1, 2 | 264 | |||
BC136515.1 | - | 1, 2 | 295 | 9020505 | ||
BC136516.1 | - | 1, 2 | 294 | 9020506 |
Set Membership: |
G-protein coupled receptor |