Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030647.1 | NP_085150.1 | 17 | 2244 | ||
XM_011516587.2 | XP_011514889.1 | 11 | 1269 | |||
GenBank mRNA | AB051505.1 | - | 5 | 549 | ||
AK131497.1 | - | 17 | 2245 | |||
BX641017.1 | - | 9 | 1024 | |||
BX649115.1 | - | 12 | 1511 |