Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030647.1 | NP_085150.1 | 12 | 1547 | ||
XM_011516587.2 | XP_011514889.1 | 6 | 572 | |||
GenBank mRNA | AK131497.1 | - | 12 | 1548 | ||
BX641017.1 | - | 4 | 327 | |||
BX649115.1 | - | 7 | 814 |