Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305008.1 | NP_001291937.1 | 10 | 986 | ||
NM_001305009.1 | NP_001291938.1 | 11 | 1074 | |||
NM_001305010.1 | NP_001291939.1 | 9 | 794 | |||
NM_001305011.1 | NP_001291940.1 | 8 | 751 | |||
NM_030650.2 | NP_085153.1 | 10 | 981 | |||
NR_130941.1 | 11 | 1270 | ||||
XM_005246878.3 | XP_005246935.1 | 10 | 917 | |||
XM_005246879.4 | XP_005246936.1 | 10 | 928 | |||
XM_005246880.3 | XP_005246937.1 | 6 | 618 | |||
XM_006712783.2 | XP_006712846.1 | 10 | 1123 | |||
XM_011511945.2 | XP_011510247.1 | 11 | 1332 | |||
XM_017005055.1 | XP_016860544.1 | 6 | 526 | |||
GenBank mRNA | AB051502.1 | - | 10 | 887 | ||
AK056532.1 | - | 10 | 904 | |||
AK301947.1 | - | 11 | 1217 | |||
AK302833.1 | - | 9 | 728 | |||
AL832947.1 | - | 7 | 644 | |||
BC105132.1 | - | 10 | 911 | 8144135 | ||
BC105134.1 | - | 10 | 911 | 8144137 | ||
BC110329.1 | - | 8 | 680 | 5732809 | ||
BC143681.1 | - | 11 | 1004 | 9052195 | ||
BC143683.1 | - | 9 | 724 | |||
CR936742.1 | - | 10 | 930 |