Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001911.2 | NP_001902.1 | 3, 4 | 370 | ||
XM_011536499.1 | XP_011534801.1 | 3, 4 | 390 | |||
GenBank mRNA | AK225914.1 | - | 2, 3 | 817 | ||
BC014460.1 | - | 3, 4 | 370 | 4859911 | ||
CR456807.1 | - | 3, 4 | 333 | |||
CR541704.1 | - | 3, 4 | 333 | |||
M16117.1 | - | 3, 4 | 341 |