Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030777.3 | NP_110404.1 | 2 | 657 | ||
XM_011529060.2 | XP_011527362.1 | 2 | 740 | |||
XM_011529061.2 | XP_011527363.1 | 2 | 2297 | |||
XM_011529062.2 | XP_011527364.1 | 2 | 751 | |||
XM_011529063.2 | XP_011527365.1 | 2 | 750 | |||
XM_011529064.2 | XP_011527366.1 | 2 | 752 | |||
XM_011529065.2 | XP_011527367.1 | 2 | 747 | |||
XM_017028087.1 | XP_016883576.1 | 2 | 650 | |||
GenBank mRNA | AF248053.1 | - | 2 | 810 | ||
AF321240.1 | - | 2 | 612 | |||
AK055548.1 | - | 2 | 657 | |||
AK290961.1 | - | 2 | 648 | |||
AK315269.1 | - | 2 | 648 | |||
AL137188.3 | - | 1 | 568 | |||
BC101657.1 | - | 2 | 656 | 8069163 | ||
BC113423.1 | - | 2 | 656 | 8322475 |