Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000396.3 | NP_000387.1 | 2 | 275 | ||
GenBank mRNA | AK130134.1 | - | 2 | 94 | ||
AK308229.1 | - | 2 | 144 | |||
AK312880.1 | - | 2 | 92 | |||
AU118043.1 | - | 2 | 172 | |||
AU119985.1 | - | 2 | 170 | |||
AY429530.1 | - | 2 | 84 | |||
BC016058.1 | - | 2 | 157 | 4860926 | ||
CR541675.1 | - | 1 | 46 | |||
DA251123.1 | - | 2 | 275 | |||
S79895.1 | - | 2 | 187 | |||
U13665.1 | - | 2 | 151 | |||
U20280.1 | - | 2 | 64 |