Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030785.3 | NP_110412.1 | 3 | 1375 | ||
XM_011527351.1 | XP_011525653.1 | 3 | 1351 | |||
GenBank mRNA | AK093068.1 | - | 2 | 645 | ||
AK098837.1 | - | 3 | 1320 | |||
AK223347.1 | - | 3 | 1320 | |||
AL136761.1 | - | 3 | 1347 | |||
BC057785.1 | - | 3 | 1203 | |||
BC136351.1 | - | 3 | 1256 | 9020338 | ||
BC136352.1 | - | 3 | 1256 | 9020339 |